Please login to your online account to display your discounted pricing

Mouse anti-CD74, Clone: SPM523, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23294710ML

 View all options for this product


For Research Use Only

Catalog No. NBP23294710


Description & Specifications


Antigen CD74
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM523
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P044233
Host Species Mouse
Immunogen SU-DHL-4 lymphoma cells
Isotype IgG1, κ
Purification Method Purified
Quantity 1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 972
Reconstitution Protein A purified