Please login to your online account to display your discounted pricing

Mouse anti-CD74, Clone: SPM523, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477801

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Gene ID (Entrez) 972
Antigen CD74
Gene Accession No. P044233
Host Species Mouse
Immunogen SU-DHL-4 lymphoma cells
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone SPM523
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal