Please login to your online account to display your discounted pricing

Mouse anti-CD74, Clone: BU45, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23456801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23456801

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Format Purified
Conjugate Unlabeled
Isotype IgG1, κ
Reconstitution Protein A purified
Host Species Mouse
Gene ID (Entrez) 972
Primary or Secondary Primary
Antigen CD74
Regulatory Status RUO
Purification Method Purified
Gene Accession No. P044233
Clone BU45
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Immunogen B lymphoblastoid cell line HFB1
Concentration 1mg/mL
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.