Please login to your online account to display your discounted pricing

Mouse anti-CD7 (p Ser123, p Ser124, p ser123, p ser124), Clone: 4H9, FITC, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen CD7 (p Ser123, p Ser124, p ser123, p ser124)
Clone 4H9
Immunogen The immunogen for this antibody is CD7.
Gene ID (Entrez) 924
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200090

 
novus biologicals canada
NBP200090100TESTS
100 Tests Each for $296.00

NBP20009025

 
novus biologicals canada
NBP20009025TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD7 (p Ser123, p Ser124, p ser123, p ser124)
Clone 4H9
Immunogen The immunogen for this antibody is CD7.
Gene ID (Entrez) 924
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry