Please login to your online account to display your discounted pricing

Mouse anti-CD7, Clone: 124-1D1, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen CD7
Clone 124-1D1
Immunogen The immunogen for this antibody was CD7.
Gene ID (Entrez) 924
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14345101

 
novus biologicals canada
NBP14345101MG
0.1mg Each for $285.00

NBP14345103

 
novus biologicals canada
NBP1434510025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen CD7
Clone 124-1D1
Immunogen The immunogen for this antibody was CD7.
Gene ID (Entrez) 924
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry