Please login to your online account to display your discounted pricing

Mouse anti-CD7, Clone: 124-1D1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14345101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14345101

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Conjugate Unlabeled
Isotype IgG1
Gene ID (Entrez) 924
Clone 124-1D1
Quantity 0.1mg
Formulation PBS [pH 7.2]
Immunogen The immunogen for this antibody was CD7.
Cross Reactivity Human
Antigen CD7
Format Affinity Purified
Purification Method Affinity Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary