Please login to your online account to display your discounted pricing

Mouse anti-CD7, Clone: 124-1D1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434510025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14345103

  • / Each
 

Description & Specifications

Specifications

Quantity 0.025mg
Format Affinity Purified
Formulation PBS [pH 7.2]
Regulatory Status RUO
Antigen CD7
Clone 124-1D1
Isotype IgG1
Gene ID (Entrez) 924
Applications Flow Cytometry
Cross Reactivity Human
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Host Species Mouse
Immunogen The immunogen for this antibody was CD7.