Please login to your online account to display your discounted pricing

Mouse anti-CD7, Clone: 124-1D1, Biotin, Novus Biologicals™

$98.00 - $197.00

Specifications

Antigen CD7
Clone 124-1D1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD7.
Gene ID (Entrez) 924
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14368501

 
novus biologicals canada
NBP14368501MG
0.1mg Each for $197.00

NBP14368503

 
novus biologicals canada
NBP1436850025MG
0.025mg Each for $98.00
Description & Specifications

Specifications

Antigen CD7
Clone 124-1D1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD7.
Gene ID (Entrez) 924
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry