Please login to your online account to display your discounted pricing

Mouse anti-CD68/SR-D1, Clone: SPM130, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CD68/SR-D1
Clone SPM130
Concentration 0.2mg/mL
Immunogen Subcellular fraction of human alveolar macrophages
Gene Accession No. P34810
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23283101

 
novus biologicals canada
NBP23283101ML
0.1mL Each for $139.00

NBP23283110

 
novus biologicals canada
NBP23283110ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen CD68/SR-D1
Clone SPM130
Concentration 0.2mg/mL
Immunogen Subcellular fraction of human alveolar macrophages
Gene Accession No. P34810
Gene ID (Entrez) 968
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting