Please login to your online account to display your discounted pricing

Mouse anti-CD68/SR-D1, Clone: SPM130, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23283110ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23283110

  • / Each
 

Description & Specifications

Specifications

Antigen CD68/SR-D1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM130
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P34810
Host Species Mouse
Immunogen Subcellular fraction of human alveolar macrophages
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 968
Test Specificity Does not react to Canine, Chicken, or Porcine.
Reconstitution Protein A purified