Please login to your online account to display your discounted pricing

Mouse anti-CD68/SR-D1, Clone: SPM130, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473602

  • / Each
 

Description & Specifications

Specifications

Antigen CD68/SR-D1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Subcellular fraction of human alveolar macrophages
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Quantity 0.2mg
Primary or Secondary Primary
Test Specificity This antibody recognizes a glycoprotein of 110kDa, which is identified as CD68. Does not react with Canine, Chicken or Porcine.
Conjugate Unlabeled
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Gene Accession No. P34810
Gene ID (Entrez) 968
Reconstitution Protein A purified
Clone SPM130