Please login to your online account to display your discounted pricing

Mouse anti-CD68/SR-D1, Clone: SPM130, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23473601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23473601

  • / Each
 

Description & Specifications

Specifications

Antigen CD68/SR-D1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM130
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P34810
Host Species Murine
Immunogen Subcellular fraction of human alveolar macrophages
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 968
Test Specificity This antibody recognizes a glycoprotein of 110kDa, which is identified as CD68. Does not react with Canine, Chicken or Porcine.
Reconstitution Protein A purified
Target Species Feline
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat