Please login to your online account to display your discounted pricing

Mouse anti-CD68/SR-D1, Clone: KP1 + C68/684, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23432901ML

 View all options for this product


For Research Use Only

Catalog No. NBP23432901


Description & Specifications


Antigen CD68/SR-D1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone KP1 + C68/684
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P34810
Host Species Mouse
Immunogen Subcellular fraction of human alveolar macrophages
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 968
Reconstitution Protein A purified