missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD68/SR-D1, Clone: KP1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD68/SR-D1
Clone KP1
Concentration 1mg/mL
Immunogen Subcellular fraction of human alveolar macrophages
Gene Accession No. P34810
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
NBP23466101 Novus Biologicals Canada
NBP23466101MG
0.1mg Each for $384.00
NBP23466102 Novus Biologicals Canada
NBP23466102MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD68/SR-D1
1mg/mL
P34810
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
KP1
Subcellular fraction of human alveolar macrophages
968
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot