Please login to your online account to display your discounted pricing

Mouse anti-CD68/SR-D1, Clone: KP1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466102

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Primary or Secondary Primary
Quantity 0.2mg
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Immunogen Subcellular fraction of human alveolar macrophages
Gene Accession No. P34810
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Antigen CD68/SR-D1
Reconstitution Protein A purified
Format Purified
Gene ID (Entrez) 968
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Purification Method Purified
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Does not react to Canine, Chicken or Porcine.
Clone KP1