Please login to your online account to display your discounted pricing

Mouse anti-CD68/SR-D1, Clone: KP1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466101

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 968
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Test Specificity Does not react to Canine, Chicken or Porcine.
Format Purified
Host Species Mouse
Quantity 0.1mg
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG1, κ
Antigen CD68/SR-D1
Clone KP1
Regulatory Status RUO
Immunogen Subcellular fraction of human alveolar macrophages
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Reconstitution Protein A purified
Gene Accession No. P34810