Please login to your online account to display your discounted pricing

Mouse anti-CD63, Clone: SPM524, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CD63
Clone SPM524
Concentration 0.2mg/mL
Immunogen Smooth plasma membrane fraction of MeWo cells
Gene Accession No. P08962
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23294301

 
novus biologicals canada
NBP23294301ML
0.1mL Each for $139.00

NBP23294310

 
novus biologicals canada
NBP23294310ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen CD63
Clone SPM524
Concentration 0.2mg/mL
Immunogen Smooth plasma membrane fraction of MeWo cells
Gene Accession No. P08962
Gene ID (Entrez) 967
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting