Please login to your online account to display your discounted pricing

Mouse anti-CD63, Clone: SPM524, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477902

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.2mg
Cross Reactivity Human
Cross Reactivity Mouse
Antigen CD63
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P08962
Immunogen Smooth plasma membrane fraction of MeWo cells
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity The tetraspanins are integral membrane proteins expressed on cell surface and granular membranes of hematopoietic cells and are components of multi-molecular complexes with specific integrins. The tetraspanin CD63 is a lysosomal membrane glycoprotein that translocates to the plasma membrane after platelet activation. CD63 is expressed on activated platelets, monocytes and macrophages, and is weakly expressed on granulocytes, T cell and B cells. It is located on the basophilic granule membranes and on the plasma membranes of lymphocytes and granulocytes. CD63 is a member of the TM4 superfamily of leukocyte glycoproteins that includes CD9, CD37 and CD53, which contain four transmembrane regions. CD63 may play a role in phagocytic and intracellular lysosome-phagosome fusion events. CD63 deficiency is associated with Hermansky-Pudlak syndrome and is strongly expressed during the early stages of melanoma progression.
Concentration 1mg/mL
Host Species Mouse
Purification Method Purified
Gene ID (Entrez) 967
Reconstitution Protein A purified
Clone SPM524
Conjugate Unlabeled