missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD63, Clone: MX-49.129.5, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD63
Clone MX-49.129.5
Concentration 1mg/mL
Immunogen Full length CD63 of human origin
Gene Accession No. P08962
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23468901 Novus Biologicals Canada
NBP23468901MG
0.1mg Each for $384.00
NBP23468902 Novus Biologicals Canada
NBP23468902MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD63
1mg/mL
P08962
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
MX-49.129.5
Full length CD63 of human origin
967
Monoclonal
Store at -20 to -80°C. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot