Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: SPM547, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CD6
Clone SPM547
Concentration 0.2mg/mL
Immunogen Human recombinant CD6 protein
Gene Accession No. P30203
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23282801

 
novus biologicals canada
NBP23282801ML
0.1mL Each for $139.00

NBP23282810

 
novus biologicals canada
NBP23282810ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen CD6
Clone SPM547
Concentration 0.2mg/mL
Immunogen Human recombinant CD6 protein
Gene Accession No. P30203
Gene ID (Entrez) 923
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting