Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: SPM547, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23282810ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23282810

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD6
Cross Reactivity Human
Regulatory Status RUO
Formulation PBS with 0.05% BSA. Protein G purified.
Conjugate Unlabeled
Gene ID (Entrez) 923
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Purification Method Purified
Clone SPM547
Primary or Secondary Primary
Quantity 1.0mL
Monoclonal or Polyclonal Monoclonal
Isotype IgG1
Gene Accession No. P30203
Concentration 0.2mg/mL
Immunogen Human recombinant CD6 protein
Format Purified