Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: SPM547, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23282801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23282801

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 923
Concentration 0.2mg/mL
Conjugate Unlabeled
Antigen CD6
Format Purified
Immunogen Human recombinant CD6 protein
Isotype IgG1
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene Accession No. P30203
Clone SPM547