Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: SPM547, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD6
Clone SPM547
Concentration 1mg/mL
Immunogen Human recombinant CD6 protein
Gene Accession No. P30203
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23478901

 
novus biologicals canada
NBP23478901MG
0.1mg Each for $384.00

NBP23478902

 
novus biologicals canada
NBP23478902MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen CD6
Clone SPM547
Concentration 1mg/mL
Immunogen Human recombinant CD6 protein
Gene Accession No. P30203
Gene ID (Entrez) 923
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting