missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: C6/372, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD6
Clone C6/372
Concentration 1mg/mL
Immunogen Human recombinant CD6 protein
Gene Accession No. P30203
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23458402 Novus Biologicals Canada
NBP23458402MG
0.2mg Each for $527.00
NBP23458401 Novus Biologicals Canada
NBP23458401MG
0.1mg Each for $384.00
Specifications Description & Specifications

Specifications

CD6
1mg/mL
P30203
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
C6/372
Human recombinant CD6 protein
923
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
ELISA,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot