Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: C6/372, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458401

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Cross Reactivity Human
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Format Purified
Gene ID (Entrez) 923
Immunogen Human recombinant CD6 protein
Antigen CD6
Clone C6/372
Isotype IgG1
Primary or Secondary Primary
Gene Accession No. P30203
Regulatory Status RUO
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Quantity 0.1mg
Purification Method Purified