Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: C6/372 + 3F7B5, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CD6
Clone C6/372 + 3F7B5
Concentration 0.2mg/mL
Immunogen Human recombinant CD6 protein (C6/372); Human rheumatoid synovial T cell line ST-1 (3F7B5)
Gene Accession No. P30203
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23431701

 
novus biologicals canada
NBP23431701ML
0.1mL Each for $139.00

NBP23431705

 
novus biologicals canada
NBP23431705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CD6
Clone C6/372 + 3F7B5
Concentration 0.2mg/mL
Immunogen Human recombinant CD6 protein (C6/372); Human rheumatoid synovial T cell line ST-1 (3F7B5)
Gene Accession No. P30203
Gene ID (Entrez) 923
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting