Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: C6/372 + 3F7B5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23431701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23431701

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD6
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P30203
Host Species Mouse
Isotype IgG1
Quantity 0.1mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 923
Conjugate Unlabeled
Cross Reactivity Human
Immunogen Human recombinant CD6 protein (C6/372); Human rheumatoid synovial T cell line ST-1 (3F7B5)
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone C6/372 + 3F7B5
Concentration 0.2mg/mL