Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: C6/372 + 3F7B5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458501

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Antigen CD6
Format Purified
Gene Accession No. P30203
Host Species Mouse
Immunogen Human recombinant CD6 protein (C6/372); Human rheumatoid synovial T cell line ST-1 (3F7B5)
Isotype IgG1
Purification Method Purified
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 923
Reconstitution Protein A purified
Clone C6/372 + 3F7B5
Concentration 1mg/mL
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal