Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: C6/372 + 3F7B5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458502

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human recombinant CD6 protein (C6/372); Human rheumatoid synovial T cell line ST-1 (3F7B5)
Purification Method Purified
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 923
Antigen CD6
Gene Accession No. P30203
Isotype IgG1
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone C6/372 + 3F7B5
Concentration 1mg/mL
Conjugate Unlabeled