missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: 3F7B5, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD6
Clone 3F7B5
Concentration 1mg/mL
Immunogen Human rheumatoid synovial T cell line ST-1
Gene Accession No. P30203
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23453701 Novus Biologicals Canada
NBP23453701MG
0.1mg Each for $384.00
NBP23453702 Novus Biologicals Canada
NBP23453702MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD6
1mg/mL
P30203
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
3F7B5
Human rheumatoid synovial T cell line ST-1
923
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
ELISA,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot