Please login to your online account to display your discounted pricing

Mouse anti-CD6, Clone: 3F7B5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23453702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23453702

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen CD6
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P30203
Host Species Mouse
Immunogen Human rheumatoid synovial T cell line ST-1
Isotype IgG1
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 923
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Primary or Secondary Primary
Reconstitution Protein A purified
Clone 3F7B5