Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: UCHT2, PE, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen CD5
Clone UCHT2
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD5.
Gene ID (Entrez) 921
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143901

 
novus biologicals canada
NBP143901100TESTS
100 Tests Each for $296.00

NBP14390125

 
novus biologicals canada
NBP14390125TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD5
Clone UCHT2
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD5.
Gene ID (Entrez) 921
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry