Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: UCHT2, PE-Cy7, Novus Biologicals™

$197.00 - $362.00

Specifications

Antigen CD5
Clone UCHT2
Immunogen The immunogen for this antibody was CD5.
Gene ID (Entrez) 921
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP142972

 
novus biologicals canada
NBP142972100TESTS
100 Tests Each for $362.00

NBP14297225

 
novus biologicals canada
NBP14297225TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen CD5
Clone UCHT2
Immunogen The immunogen for this antibody was CD5.
Gene ID (Entrez) 921
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE-Cy7
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry