missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: UCHT2, Novus Biologicals™

$219.00 - $274.00

Specifications

Antigen CD5
Clone UCHT2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD5.
Gene ID (Entrez) 921
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14345203 Novus Biologicals Canada
NBP1434520025MG
0.025mg Each for $219.00
NBP14345201 Novus Biologicals Canada
NBP14345201MG
0.1mg Each for $274.00
Specifications Description & Specifications

Specifications

CD5
0.5mg/mL
921
Monoclonal
Store at 4°C. Do not freeze.
IgG1, κ
Affinity Purified
Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen),Western Blot
UCHT2
The immunogen for this antibody was CD5.
Aqueous buffer (contains stabilizer if necessary)
Primary
Murine
Unlabeled
Affinity Purified