Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: UCHT2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14345201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14345201

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Cross Reactivity Human
Antigen CD5
Formulation Aqueous buffer (contains stabilizer if necessary)
Format Affinity Purified
Gene ID (Entrez) 921
Immunogen The immunogen for this antibody was CD5.
Purification Method Affinity Purified
Concentration 0.5mg/mL
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Conjugate Unlabeled
Clone UCHT2
Isotype IgG1, κ
Storage Requirements Store at 4°C. Do not freeze.
Test Specificity This antibody is reactive to Human, and recognizes Primate CD 5.
Quantity 0.1mg