Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: UCHT2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434520025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14345203

  • / Each
 

Description & Specifications

Specifications

Antigen CD5
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blot
Clone UCHT2
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary)
Host Species Murine
Immunogen The immunogen for this antibody was CD5.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 921
Test Specificity This antibody is reactive to Human, and recognizes Primate CD 5.
Target Species Human