Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: UCHT2, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD5
Clone UCHT2
Immunogen The immunogen for this antibody was CD5.
Gene ID (Entrez) 921
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14368601

 
novus biologicals canada
NBP14368601MG
0.1mg Each for $252.00

NBP14368603

 
novus biologicals canada
NBP1436860025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen CD5
Clone UCHT2
Immunogen The immunogen for this antibody was CD5.
Gene ID (Entrez) 921
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)