Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: SPM546, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23478801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23478801

  • / Each
 

Description & Specifications

Specifications

Antigen CD5
Gene Accession No. P06127
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human CD5 recombinant protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 921
Reconstitution Protein A purified
Clone SPM546
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Recognizes a 67kDa transmembrane protein, which is identified as CD5. Anti-CD5 does not react with granulocytes or monocytes.