Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: L17F12, FITC, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD5
Clone L17F12
Immunogen The immunogen for this antibody was CD5.
Gene ID (Entrez) 921
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP144041

 
novus biologicals canada
NBP144041100TESTS
100 Tests Each for $252.00

NBP14404125

 
novus biologicals canada
NBP14404125TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CD5
Clone L17F12
Immunogen The immunogen for this antibody was CD5.
Gene ID (Entrez) 921
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry