Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: CRIS-1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD5
Clone CRIS-1
Concentration 1mg/mL
Immunogen Stimulated human leukocytes
Gene Accession No. P06127
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23459802

 
novus biologicals canada
NBP23459802MG
0.2mg Each for $527.00

NBP23459801

 
novus biologicals canada
NBP23459801MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen CD5
Clone CRIS-1
Concentration 1mg/mL
Immunogen Stimulated human leukocytes
Gene Accession No. P06127
Gene ID (Entrez) 921
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation