Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: CRIS-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23459801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23459801

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rhesus Ma
Immunogen Stimulated human leukocytes
Isotype IgG2a, κ
Conjugate Unlabeled
Primary or Secondary Primary
Gene Accession No. P06127
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg
Format Purified
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen CD5
Clone CRIS-1
Concentration 1mg/mL
Gene ID (Entrez) 921
Regulatory Status RUO