Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: C5/473+CD5/54/F6, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CD5
Clone C5/473+CD5/54/F6
Concentration 0.2mg/mL
Immunogen Human CD5 recombinant protein (C5/473); A synthetic peptide from the intracellular region of human CD5 (CD5/54/F6)
Gene Accession No. P06127
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23431601

 
novus biologicals canada
NBP23431601ML
0.1mL Each for $139.00

NBP23431605

 
novus biologicals canada
NBP23431605ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CD5
Clone C5/473+CD5/54/F6
Concentration 0.2mg/mL
Immunogen Human CD5 recombinant protein (C5/473); A synthetic peptide from the intracellular region of human CD5 (CD5/54/F6)
Gene Accession No. P06127
Gene ID (Entrez) 921
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting