Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: C5/473+CD5/54/F6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23431601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23431601

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Antigen CD5
Formulation PBS with 0.05% BSA.
Gene Accession No. P06127
Immunogen Human CD5 recombinant protein (C5/473); A synthetic peptide from the intracellular region of human CD5 (CD5/54/F6)
Isotype IgG1, κ
Purification Method Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone C5/473+CD5/54/F6
Conjugate Unlabeled
Format Purified
Quantity 0.1mL
Gene ID (Entrez) 921
Concentration 0.2mg/mL