missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: C5/473+CD5/54/F6, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD5
Clone C5/473+CD5/54/F6
Concentration 1mg/mL
Immunogen Human CD5 recombinant protein (C5/473); A synthetic peptide from the intracellular region of human CD5 (CD5/54/F6)
Gene Accession No. P06127
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23458301 Novus Biologicals Canada
NBP23458301MG
0.1mg Each for $384.00
NBP23458302 Novus Biologicals Canada
NBP23458302MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CD5
1mg/mL
P06127
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
C5/473+CD5/54/F6
Human CD5 recombinant protein (C5/473); A synthetic peptide from the intracellular region of human CD5 (CD5/54/F6)
921
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot