Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: C5/473+CD5/54/F6, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458302

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Gene ID (Entrez) 921
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.2mg
Gene Accession No. P06127
Clone C5/473+CD5/54/F6
Cross Reactivity Human
Immunogen Human CD5 recombinant protein (C5/473); A synthetic peptide from the intracellular region of human CD5 (CD5/54/F6)
Conjugate Unlabeled
Concentration 1mg/mL
Format Purified
Primary or Secondary Primary
Purification Method Purified
Isotype IgG1, κ
Reconstitution Protein A purified
Antigen CD5
Host Species Mouse