Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: C5/473+CD5/54/F6, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458301

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Regulatory Status RUO
Gene ID (Entrez) 921
Cross Reactivity Human
Antigen CD5
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P06127
Host Species Mouse
Immunogen Human CD5 recombinant protein (C5/473); A synthetic peptide from the intracellular region of human CD5 (CD5/54/F6)
Isotype IgG1, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone C5/473+CD5/54/F6
Concentration 1mg/mL
Conjugate Unlabeled