Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: C5/473, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CD5
Clone C5/473
Concentration 1mg/mL
Immunogen Human CD5 recombinant protein
Gene Accession No. P06127
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23458201

 
novus biologicals canada
NBP23458201MG
0.1mg Each for $384.00

NBP23458202

 
novus biologicals canada
NBP23458202MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen CD5
Clone C5/473
Concentration 1mg/mL
Immunogen Human CD5 recombinant protein
Gene Accession No. P06127
Gene ID (Entrez) 921
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting