Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: C5/473, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458201

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Immunogen Human CD5 recombinant protein
Quantity 0.1mg
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Clone C5/473
Gene Accession No. P06127
Gene ID (Entrez) 921
Conjugate Unlabeled
Regulatory Status RUO
Reconstitution Protein A purified
Antigen CD5
Host Species Mouse
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Isotype IgG1, κ
Format Purified
Purification Method Purified
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.