Please login to your online account to display your discounted pricing

Mouse anti-CD5, Clone: C5/473, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23458202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23458202

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.2mg
Gene ID (Entrez) 921
Concentration 1mg/mL
Cross Reactivity Human
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P06127
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Isotype IgG1, κ
Conjugate Unlabeled
Immunogen Human CD5 recombinant protein
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Purification Method Purified
Clone C5/473
Antigen CD5
Regulatory Status RUO