missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD46, Clone: 122.2, Novus Biologicals™

$142.00 - $472.00

Specifications

Antigen CD46
Clone 122.2
Concentration 0.2mg/mL
Immunogen A recombinant protein fragment specific for human CD46 was used as the immunogen.
Gene Accession No. NP_002380
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP2252222M Novus Biologicals Canada
NBP22522202MG
0.2mg Each for $472.00
NBP22522202 Novus Biologicals Canada
NBP225222002MG
0.02mg Each for $142.00
Specifications Description & Specifications

Specifications

CD46
0.2mg/mL
NP_002380
PBS with 0.05% BSA. Protein G purified.
Primary
Murine
Unlabeled
Purified
122.2
A recombinant protein fragment specific for human CD46 was used as the immunogen.
4179
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry